Assay Details
| Gene Symbol: OTUD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018054897.1 | XP_017910386.1 | 7 - 8 | 1154 | 71 | |
| XM_018054901.1 | XP_017910390.1 | 6 - 7 | 1032 | 71 | ||
| GenBank mRNA | KA307836.1 | - | 1 - 2 | 2281 | 71 | |
More Information
Set Membership: |
|

OTUD3
Genomic Map