Assay Details
| Gene Symbol: LOC102170378 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005678259.2 | XP_005678316.1 | 11 - 12 | 1241 | 91 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102170378| Gene Symbol: LOC102170378 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005678259.2 | XP_005678316.1 | 11 - 12 | 1241 | 91 | |
Set Membership: |
|