Assay Details
Gene Symbol: LOC102170378 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005678259.2 | XP_005678316.1 | 1 - 2 | 101 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
