Assay Details
| Gene Symbol: LOC102170378 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005678259.2 | XP_005678316.1 | 5 - 6 | 512 | 73 | |
| GenBank mRNA | JO372142.2 | - | 4 - 5 | 398 | 73 | |
More Information
Set Membership: |
|

LOC102170378
Genomic Map