Assay Details
| Gene Symbol: CRADD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005679822.3 | XP_005679879.1 | 2 - 3 | 433 | 78 | |
| GenBank mRNA | JO400657.2 | - | 1 - 2 | 215 | 78 | |
More Information
Set Membership: |
|

CRADD
Genomic Map