Assay Details
| Gene Symbol: RND1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005680013.2 | XP_005680070.1 | 4 - 5 | 563 | 58 | |
| GenBank mRNA | JO453992.2 | - | 4 - 5 | 566 | 58 | |
| JO623831.2 | - | 1 - 2 | 1068 | 58 | ||
More Information
Set Membership: |
|

RND1
Genomic Map