Assay Details
| Gene Symbol: LOC102174022 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005683119.3 | XP_005683176.2 | 25 - 26 | 2522 | 96 | |
| XM_005683121.3 | XP_005683178.2 | 25 - 26 | 2519 | 96 | ||
| XM_005683122.3 | XP_005683179.2 | 25 - 26 | 2519 | 96 | ||
| XM_005683123.3 | XP_005683180.2 | 25 - 26 | 2522 | 96 | ||
| XM_005683125.3 | XP_005683182.2 | 23 - 24 | 2312 | 96 | ||
More Information
Set Membership: |
|

LOC102174022
Genomic Map