Assay Details
| Gene Symbol: STX7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005684757.3 | XP_005684814.1 | 4 - 5 | 445 | 71 | |
| GenBank mRNA | JO581991.1 | - | 4 - 5 | 498 | 71 | |
| JO581992.1 | - | 4 - 5 | 498 | 71 | ||
More Information
Set Membership: |
|

STX7
Genomic Map