Assay Details
| Gene Symbol: IVD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018054448.1 | XP_017909937.1 | 4 - 5 | 628 | 62 | |
| XM_018054449.1 | XP_017909938.1 | 4 - 5 | 628 | 62 | ||
More Information
Set Membership: |
|

IVD
Genomic Map