Assay Details
| Gene Symbol: CDKN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005685875.3 | XP_005685932.1 | 2 - 3 | 214 | 96 | |
| XM_005685876.3 | XP_005685933.1 | 2 - 3 | 214 | 96 | ||
| XM_018054126.1 | XP_017909615.1 | 2 - 3 | 214 | 96 | ||
More Information
Set Membership: |
|

CDKN3
Genomic Map