Assay Details
| Gene Symbol: ZC2HC1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005686094.3 | XP_005686151.1 | 2 - 3 | 1436 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZC2HC1C| Gene Symbol: ZC2HC1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005686094.3 | XP_005686151.1 | 2 - 3 | 1436 | 84 | |
Set Membership: |
|