Assay Details
| Gene Symbol: CPB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005687437.3 | XP_005687494.1 | 11 - 12 | 1409 | 65 | |
| GenBank mRNA | JO421612.2 | - | 13 - 14 | 2367 | 65 | |
More Information
Set Membership: |
|

CPB2
Genomic Map