Assay Details
| Gene Symbol: BIRC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018059894.1 | XP_017915383.1 | 3 - 4 | 2401 | 62 | |
| XM_018059895.1 | XP_017915384.1 | 3 - 4 | 2401 | 62 | ||
| GenBank mRNA | JO415431.2 | - | 828 | 62 | ||
More Information
Set Membership: |
|

BIRC2
Genomic Map