Assay Details
| Gene Symbol: PLA2G3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005691644.3 | XP_005691701.2 | 2 - 3 | 786 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PLA2G3| Gene Symbol: PLA2G3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005691644.3 | XP_005691701.2 | 2 - 3 | 786 | 79 | |
Set Membership: |
|