Assay Details
| Gene Symbol: LOC102171189 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005693879.3 | XP_005693936.2 | 10 - 11 | 1093 | 91 | |
| GenBank mRNA | JO435028.2 | - | 2 - 3 | 426 | 91 | |
More Information
Set Membership: |
|

LOC102171189
Genomic Map