Assay Details
| Gene Symbol: EGLN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005695224.3 | XP_005695281.1 | 1 - 2 | 682 | 69 | |
| GenBank mRNA | JO426510.1 | - | 1 - 2 | 714 | 69 | |
More Information
Set Membership: |
|

EGLN3
Genomic Map