Assay Details
| Gene Symbol: TM2D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005698843.3 | XP_005698900.1 | 1 - 2 | 450 | 64 | |
| GenBank mRNA | JI861796.1 | - | 1 - 2 | 232 | 64 | |
| JT753549.1 | - | 6 - 7 | 1098 | 64 | ||
More Information
Set Membership: |
|

TM2D2
Genomic Map