Assay Details
| Gene Symbol: TM7SF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005699882.3 | XP_005699939.2 | 7 - 8 | 1115 | 64 | |
| GenBank mRNA | JI861829.1 | - | 7 - 8 | 890 | 64 | |
More Information
Set Membership: |
|

TM7SF2
Genomic Map