Assay Details
| Gene Symbol: ZSWIM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005709660.3 | XP_005709717.2 | 2 | 1818 | 77 | |
| XM_018057936.1 | XP_017913425.1 | 3 | 1862 | 77 | ||
More Information
Set Membership: |
|

ZSWIM1
Genomic Map