Assay Details
| Gene Symbol: LOC102173333 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JO397036.2 | - | 2 - 3 | 516 | 117 | |
| JO397037.2 | - | 2 - 3 | 516 | 117 | ||
| JO429415.2 | - | 2 - 3 | 548 | 117 | ||
| JT717547.1 | - | 2 - 3 | 491 | 117 | ||
| KA305766.1 | - | 14 - 15 | 1678 | 117 | ||
| Gene Symbol: LOC108635581 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018045824.1 | XP_017901313.1 | 2 - 3 | 671 | 117 | |
| XM_018045834.1 | XP_017901323.1 | 2 - 3 | 676 | 117 | ||
| GenBank mRNA | JT705848.1 | - | 2 - 3 | 369 | 117 | |
More Information
Set Membership: |
|

LOC102173333
Genomic Map