Assay Details
| Gene Symbol: LOC102173333 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JO371919.2 | - | 6 - 7 | 836 | 109 | |
| JO371920.2 | - | 5 - 6 | 446 | 109 | ||
| JO397036.2 | - | 6 - 7 | 916 | 109 | ||
| JO429415.2 | - | 6 - 7 | 948 | 109 | ||
| JO429432.2 | - | 6 - 7 | 945 | 109 | ||
| JO548802.2 | - | 3 - 4 | 209 | 109 | ||
| JO596232.2 | - | 8 - 9 | 984 | 109 | ||
| JO615717.2 | - | 4 - 5 | 351 | 109 | ||
| JO615718.2 | - | 4 - 5 | 351 | 109 | ||
| JT717547.1 | - | 7 - 8 | 987 | 109 | ||
| JT737734.1 | - | 2 - 3 | 128 | 109 | ||
| KA305766.1 | - | 10 - 11 | 1278 | 109 | ||
| Gene Symbol: LOC108635581 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018045824.1 | XP_017901313.1 | 6 - 7 | 1071 | 109 | |
| XM_018045830.1 | XP_017901319.1 | 6 - 7 | 1069 | 109 | ||
| XM_018045832.1 | XP_017901321.1 | 6 - 7 | 957 | 109 | ||
| XM_018045834.1 | XP_017901323.1 | 7 - 8 | 1172 | 109 | ||
| XM_018045836.1 | XP_017901325.1 | 7 - 8 | 1057 | 109 | ||
| GenBank mRNA | JT705848.1 | - | 6 - 7 | 770 | 109 | |
More Information
Set Membership: |
|

LOC102173333
Genomic Map