Assay Details
| Gene Symbol: LOC102173333 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JO371919.2 | - | 10 - 11 | 1139 | 63 | |
| JO371920.2 | - | 9 - 10 | 749 | 63 | ||
| JO397036.2 | - | 10 - 11 | 1219 | 63 | ||
| JO429415.2 | - | 10 - 11 | 1251 | 63 | ||
| JO429432.2 | - | 10 - 11 | 1248 | 63 | ||
| JO548802.2 | - | 7 - 8 | 512 | 63 | ||
| JO596232.2 | - | 4 - 5 | 681 | 63 | ||
| JO615717.2 | - | 8 - 9 | 654 | 63 | ||
| JO615718.2 | - | 8 - 9 | 654 | 63 | ||
| JT717547.1 | - | 11 - 12 | 1292 | 63 | ||
| KA305766.1 | - | 6 - 7 | 975 | 63 | ||
| Gene Symbol: LOC108635581 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018045824.1 | XP_017901313.1 | 10 - 11 | 1374 | 63 | |
| XM_018045830.1 | XP_017901319.1 | 10 - 11 | 1372 | 63 | ||
| XM_018045832.1 | XP_017901321.1 | 10 - 11 | 1260 | 63 | ||
| XM_018045834.1 | XP_017901323.1 | 11 - 12 | 1475 | 63 | ||
| XM_018045836.1 | XP_017901325.1 | 11 - 12 | 1360 | 63 | ||
| GenBank mRNA | JT705848.1 | - | 10 - 11 | 1074 | 63 | |
More Information
Set Membership: |
|

LOC102173333
Genomic Map