Assay Details
| Gene Symbol: LOC102173333 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JO371919.2 | - | 11 - 12 | 1214 | 95 | |
| JO371920.2 | - | 10 - 11 | 824 | 95 | ||
| JO397036.2 | - | 11 - 12 | 1294 | 95 | ||
| JO429415.2 | - | 11 - 12 | 1326 | 95 | ||
| JO429432.2 | - | 11 - 12 | 1323 | 95 | ||
| JO548802.2 | - | 8 - 9 | 587 | 95 | ||
| JO596232.2 | - | 3 - 4 | 606 | 95 | ||
| JO615717.2 | - | 9 - 10 | 729 | 95 | ||
| KA305766.1 | - | 5 - 6 | 900 | 95 | ||
| Gene Symbol: LOC108635581 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018045824.1 | XP_017901313.1 | 11 - 12 | 1449 | 95 | |
| XM_018045830.1 | XP_017901319.1 | 11 - 12 | 1447 | 95 | ||
| XM_018045832.1 | XP_017901321.1 | 11 - 12 | 1335 | 95 | ||
| XM_018045834.1 | XP_017901323.1 | 12 - 13 | 1550 | 95 | ||
| XM_018045836.1 | XP_017901325.1 | 12 - 13 | 1435 | 95 | ||
More Information
Set Membership: |
|

LOC102173333
Genomic Map