Assay Details
| Gene Symbol: LOC102173333 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JO371919.2 | - | 13 - 14 | 1578 | 66 | |
| JO371920.2 | - | 12 - 13 | 1188 | 66 | ||
| JO397036.2 | - | 13 - 14 | 1658 | 66 | ||
| JO429415.2 | - | 13 - 14 | 1690 | 66 | ||
| JO429432.2 | - | 13 - 14 | 1687 | 66 | ||
| JO548802.2 | - | 10 - 11 | 951 | 66 | ||
| JO596232.2 | - | 1 - 2 | 242 | 66 | ||
| JO626050.2 | - | 2 - 3 | 396 | 66 | ||
| JT717547.1 | - | 14 - 15 | 1733 | 66 | ||
| JT737735.1 | - | 1 - 2 | 164 | 66 | ||
| KA305766.1 | - | 3 - 4 | 536 | 66 | ||
| Gene Symbol: LOC108635581 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018045824.1 | XP_017901313.1 | 13 - 14 | 1813 | 66 | |
| XM_018045830.1 | XP_017901319.1 | 13 - 14 | 1811 | 66 | ||
| XM_018045832.1 | XP_017901321.1 | 13 - 14 | 1699 | 66 | ||
| XM_018045834.1 | XP_017901323.1 | 14 - 15 | 1914 | 66 | ||
| XM_018045836.1 | XP_017901325.1 | 14 - 15 | 1799 | 66 | ||
| GenBank mRNA | JT705848.1 | - | 13 - 14 | 1521 | 66 | |
More Information
Set Membership: |
|

LOC102173333
Genomic Map