Assay Details
| Gene Symbol: LOC102181858 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018051327.1 | XP_017906816.1 | 3 - 4 | 427 | 86 | |
| XM_018051328.1 | XP_017906817.1 | 3 - 4 | 426 | 86 | ||
| XM_018051329.1 | XP_017906818.1 | 3 - 4 | 368 | 86 | ||
| GenBank mRNA | JO420154.2 | - | 3 - 4 | 86 | ||
| Gene Symbol: LOC108635485 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018045602.1 | XP_017901091.1 | 1 - 2 | 448 | 86 | |
More Information
Set Membership: |
|

LOC102181858
Genomic Map