Assay Details
| Gene Symbol: MFAP3L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018051812.1 | XP_017907301.1 | 2 - 3 | 656 | 66 | |
| XM_018051813.1 | XP_017907302.1 | 2 - 3 | 652 | 66 | ||
| GenBank mRNA | JO620815.2 | - | 2 - 3 | 617 | 66 | |
More Information
Set Membership: |
|

MFAP3L
Genomic Map