Assay Details
| Gene Symbol: FAM228B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018055570.1 | XP_017911059.1 | 2 - 3 | 483 | 65 | |
| XM_018055571.1 | XP_017911060.1 | 2 - 3 | 423 | 65 | ||
| XM_018055573.1 | XP_017911062.1 | 2 - 3 | 222 | 65 | ||
| XM_018055574.1 | XP_017911063.1 | 2 - 3 | 183 | 65 | ||
| XM_018055575.1 | XP_017911064.1 | 2 - 3 | 484 | 65 | ||
| XM_018055576.1 | XP_017911065.1 | 2 - 3 | 284 | 65 | ||
| XM_018055577.1 | XP_017911066.1 | 2 - 3 | 473 | 65 | ||
| XM_018055578.1 | XP_017911067.1 | 2 - 3 | 475 | 65 | ||
| XM_018055579.1 | XP_017911068.1 | 2 - 3 | 284 | 65 | ||
More Information
Set Membership: |
|

FAM228B
Genomic Map