Assay Details
| Gene Symbol: AK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018055832.1 | XP_017911321.1 | 5 - 6 | 455 | 104 | |
| GenBank mRNA | JO643300.2 | - | 2 - 3 | 1486 | 104 | |
| JO643301.2 | - | 2 - 3 | 1486 | 104 | ||
More Information
Set Membership: |
|

AK1
Genomic Map