Assay Details
| Gene Symbol: WEE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018059526.1 | XP_017915015.1 | 2 - 3 | 1031 | 73 | |
| GenBank mRNA | JO574021.2 | - | 4 - 5 | 420 | 73 | |
| JO585743.2 | - | 9 - 10 | 2171 | 73 | ||
More Information
Set Membership: |
|

WEE1
Genomic Map