Assay Details
| Gene Symbol: TMEM131L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061620.1 | XP_017917109.1 | 22 - 23 | 2500 | 55 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM131L| Gene Symbol: TMEM131L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061620.1 | XP_017917109.1 | 22 - 23 | 2500 | 55 | |
Set Membership: |
|