Assay Details
| Gene Symbol: LOC102182159 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061235.1 | XP_017916724.1 | 7 - 8 | 1016 | 56 | |
| XM_018061236.1 | XP_017916725.1 | 5 - 6 | 692 | 56 | ||
| XM_018061237.1 | XP_017916726.1 | 4 - 5 | 486 | 56 | ||
More Information
Set Membership: |
|

LOC102182159
Genomic Map