Assay Details
| Gene Symbol: FAM76A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018056841.1 | XP_017912330.1 | 3 - 4 | 443 | 63 | |
| XM_018056843.1 | XP_017912332.1 | 3 - 4 | 411 | 63 | ||
| GenBank mRNA | JT672222.1 | - | 6 - 7 | 908 | 63 | |
More Information
Set Membership: |
|

FAM76A
Genomic Map