Assay Details
| Gene Symbol: PRICKLE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018038277.1 | XP_017893766.1 | 8 - 9 | 1948 | 54 | |
| XM_018038278.1 | XP_017893767.1 | 7 - 8 | 1987 | 54 | ||
| XM_018038279.1 | XP_017893768.1 | 7 - 8 | 2387 | 54 | ||
More Information
Set Membership: |
|

PRICKLE2
Genomic Map