Assay Details
| Gene Symbol: LOC102177581 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040760.1 | XP_017896249.1 | 14 - 15 | 2931 | 68 | |
| XM_018040761.1 | XP_017896250.1 | 14 - 15 | 2931 | 68 | ||
More Information
Set Membership: |
|

LOC102177581
Genomic Map