Assay Details
| Gene Symbol: PMF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018046288.1 | XP_017901777.1 | 4 - 5 | 592 | 57 | |
| GenBank mRNA | JO577972.1 | - | 1 - 2 | 555 | 57 | |
More Information
Set Membership: |
|

PMF1
Genomic Map