Assay Details
| Gene Symbol: FLACC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005676367.3 | XP_005676424.1 | 3 - 4 | 281 | 89 | |
| XM_005676368.3 | XP_005676425.1 | 3 - 4 | 281 | 89 | ||
More Information
Set Membership: |
|

FLACC1
Genomic Map