Assay Details
| Gene Symbol: TRIM63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005676825.2 | XP_005676882.1 | 6 - 7 | 994 | 55 | |
| GenBank mRNA | JO356699.2 | - | 4 - 5 | 1184 | 55 | |
More Information
Set Membership: |
|

TRIM63
Genomic Map