Assay Details
| Gene Symbol: PIK3R3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005678520.3 | XP_005678577.1 | 8 - 9 | 1764 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PIK3R3| Gene Symbol: PIK3R3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005678520.3 | XP_005678577.1 | 8 - 9 | 1764 | 69 | |
Set Membership: |
|