Assay Details
| Gene Symbol: DBX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018049093.1 | XP_017904582.1 | 3 - 4 | 1013 | 68 | |
| GenBank mRNA | JT652264.1 | - | 4 - 5 | 575 | 68 | |
More Information
Set Membership: |
|

DBX2
Genomic Map