Assay Details
| Gene Symbol: NPFFR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005681743.3 | XP_005681800.2 | 3 - 4 | 574 | 91 | |
| GenBank mRNA | JT622839.1 | - | 1 - 2 | 89 | 91 | |
More Information
Set Membership: |
|

NPFFR2
Genomic Map