Assay Details
| Gene Symbol: C8H9orf131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005683929.2 | XP_005683986.2 | 1 - 2 | 333 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C8H9orf131| Gene Symbol: C8H9orf131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005683929.2 | XP_005683986.2 | 1 - 2 | 333 | 76 | |
Set Membership: |
|