Assay Details
| Gene Symbol: LOC102182036 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005684359.3 | XP_005684416.2 | 2 - 3 | 1780 | 57 | |
| XM_005684361.3 | XP_005684418.2 | 2 - 3 | 1785 | 57 | ||
More Information
Set Membership: |
|

LOC102182036
Genomic Map