Assay Details
| Gene Symbol: LOC102182036 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005684359.3 | XP_005684416.2 | 5 - 6 | 1928 | 109 | |
| XM_005684360.3 | XP_005684417.2 | 4 - 5 | 1867 | 109 | ||
More Information
Set Membership: |
|

LOC102182036
Genomic Map