Assay Details
| Gene Symbol: SYNCRIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018053167.1 | XP_017908656.1 | 4 - 5 | 909 | 64 | |
| XM_018053169.1 | XP_017908658.1 | 4 - 5 | 490 | 64 | ||
| XM_018053170.1 | XP_017908659.1 | 4 - 5 | 909 | 64 | ||
| XM_018053171.1 | XP_017908660.1 | 4 - 5 | 909 | 64 | ||
| XM_018053172.1 | XP_017908661.1 | 3 - 4 | 783 | 64 | ||
| XM_018053173.1 | XP_017908662.1 | 3 - 4 | 348 | 64 | ||
| GenBank mRNA | JO409479.2 | - | 8 - 9 | 1580 | 64 | |
| JO427781.1 | - | 8 - 9 | 1674 | 64 | ||
More Information
Set Membership: |
|

SYNCRIP
Genomic Map