Assay Details
| Gene Symbol: TPRKB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005686359.3 | XP_005686416.1 | 4 - 5 | 585 | 95 | |
| XM_005686360.3 | XP_005686417.1 | 4 - 5 | 704 | 95 | ||
| XM_005686361.3 | XP_005686418.1 | 4 - 5 | 1186 | 95 | ||
| XM_005686363.3 | XP_005686420.1 | 4 - 5 | 575 | 95 | ||
| XM_005686364.3 | XP_005686421.1 | 4 - 5 | 1183 | 95 | ||
| XM_005686365.3 | XP_005686422.1 | 4 - 5 | 701 | 95 | ||
| XM_005686367.3 | XP_005686424.1 | 4 - 5 | 680 | 95 | ||
| XM_005686368.3 | XP_005686425.1 | 4 - 5 | 679 | 95 | ||
| XM_018055030.1 | XP_017910519.1 | 4 - 5 | 573 | 95 | ||
More Information
Set Membership: |
|

TPRKB
Genomic Map