Assay Details
| Gene Symbol: ZNF706 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018058315.1 | XP_017913804.1 | 3 - 4 | 259 | 80 | |
| XM_018058317.1 | XP_017913806.1 | 3 - 4 | 520 | 80 | ||
More Information
Set Membership: |
|

ZNF706
Genomic Map