Assay Details
| Gene Symbol: DYNLRB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005691820.3 | XP_005691877.1 | 2 - 3 | 183 | 95 | |
| GenBank mRNA | JO636996.2 | - | 2 - 3 | 217 | 95 | |
More Information
Set Membership: |
|

DYNLRB2
Genomic Map