Assay Details
| Gene Symbol: WRAP53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005693531.3 | XP_005693588.1 | 8 - 9 | 1519 | 62 | |
| GenBank mRNA | JO594735.2 | - | 2 - 3 | 373 | 62 | |
More Information
Set Membership: |
|

WRAP53
Genomic Map