Assay Details
| Gene Symbol: CDK5RAP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005694286.3 | XP_005694343.3 | 6 - 7 | 704 | 64 | |
| GenBank mRNA | JO374098.2 | - | 4 - 5 | 476 | 64 | |
More Information
Set Membership: |
|

CDK5RAP3
Genomic Map