Assay Details
| Gene Symbol: HEMK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005695916.3 | XP_005695973.1 | 6 - 7 | 1159 | 61 | |
| XM_005695917.3 | XP_005695974.1 | 6 - 7 | 1161 | 61 | ||
| GenBank mRNA | JO502594.2 | - | 5 - 6 | 1027 | 61 | |
More Information
Set Membership: |
|

HEMK1
Genomic Map